Nalgene™ Narrow-Mouth HDPE Bottles with Closure: Shrink-Wrapped Trays
Nalgene™ Narrow-Mouth HDPE Bottles with Closure: Shrink-Wrapped Trays
Actual product may vary
Thermo Scientific™

Nalgene™ Narrow-Mouth HDPE Bottles with Closure: Shrink-Wrapped Trays

Easily handle these narrow-mouth HDPE bottles that are ready to feed into filling lines for various reagents, and conveniently packaged in shrink-wrap trays.
Have Questions?
Change viewbuttonViewtableView
Capacity (Metric):
30 mL
60 mL
250 mL
Closure Size:
View all
20-415 mm
Catalog number 322089-0001
Price (USD)/ Case of 1050
857.44
Online exclusive
932.00 
Save 74.56 (8%)
-
Add to cart
Capacity (Metric):
30 mL
Closure Size:
20-415 mm
Price (USD)/ Case of 1050
857.44
Online exclusive
932.00 
Save 74.56 (8%)
Add to cart
Nalgene™ Narrow-Mouth HDPE Bottles with Closure: Shrink-Wrapped Trays
Catalog number322089-0001
Price (USD)/ Case of 1050
857.44
Online exclusive
932.00 
Save 74.56 (8%)
-
Add to cart
Easily handle Thermo Scientific™ Nalgene™ Narrow-Mouth HDPE Bottles, conveniently packaged in rigid, lint-free shrink-wrap trays and ready to feed into filling lines for various reagents. High density polyethylene bottles include polypropylene closures and rigid, lint-free trays.
Features:
  • Excellent impact and chemical resistance
  • Include opaque amber polypropylene closures
  • Translucent amber bottles provide light protection with the ability to see liquid level
  • Leakproof† when used with Nalgene closures

Ordering Information:

Bottles and closures are packed in separate polybags within the same carton.

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for two minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

A sample product certificate providing Cytotoxicity, USP Class VI, FDA Compliance—21 CFR and other regulatory compliance information is available or can be requested from our Certificates of Compliance Database

Specifications
Capacity1 oz., 30 mL
Capacity (Metric)30 mL
Capacity (English)1 oz.
ColorNatural
DescriptionNarrow-Mouth Packaging Bottles with Closures
Diameter (Metric) Inner Neck14 mm
MaterialHDPE
No. per Case1050
No. per Pack70
Closure ColorNatural
Neck Finish20 mm
Certifications/ComplianceManufactured in compliance with ISO 13485:2016 quality requirements. Diagnostic Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI.
Closure IncludedYes
GraduatedNo
MouthNarrow
ShapeRound
Height (Metric)61 mm
Product TypeNarrow Mouth Bottle
Width (English)1.35 in.
Height (English)2.4 in.
Closure MaterialPolypropylene
Closure Size20-415 mm
Diameter (Metric)34 mm
Closure TypeScrew (Bulk Packed, Unassembled)
Through the Closure DispensingNo
Unit SizeCase of 1050