(-)-Caryophyllene oxide, 95%, Thermo Scientific Chemicals
(-)-Caryophyllene oxide, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

(-)-Caryophyllene oxide, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
250 g
Packaging:
Glass bottle
Catalog number 325080500
Price (USD)/ Each
61.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
72.90 
Save 11.25 (15%)
-
Add to cart
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
61.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
72.90 
Save 11.25 (15%)
Add to cart
(-)-Caryophyllene oxide, 95%, Thermo Scientific Chemicals
Catalog number325080500
Price (USD)/ Each
61.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
72.90 
Save 11.25 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Name4,12,12-trimethyl-9-methylidene-5-oxatricyclo[8.2.0.0⁴,⁶]dodecane
Molecular FormulaC15H24O
InChI KeyNVEQFIOZRFFVFW-UHFFFAOYNA-N
SMILESCC12CCC3C(CC3(C)C)C(=C)CCC1O2
Molecular Weight (g/mol)220.36
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Melting point55°C to 64°C
Appearance (Form)Crystalline powder
Appearance (Color)White
GC>=94 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only