(-)-Caryophyllene oxide, 95%, Thermo Scientific Chemicals
(-)-Caryophyllene oxide, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

(-)-Caryophyllene oxide, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
10 g
50 g
Packaging:
Glass bottle
Catalog number 325082500
Price (USD)/ Each
-
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1139-30-6
IUPAC Name4,12,12-trimethyl-9-methylidene-5-oxatricyclo[8.2.0.0⁴,⁶]dodecane
Molecular FormulaC15H24O
InChI KeyNVEQFIOZRFFVFW-UHFFFAOYNA-N
SMILESCC12CCC3C(CC3(C)C)C(=C)CCC1O2
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=94 %
Specific optical rotation-55° to -75° (20°C, 589 nm) (c=10, EtOH)
Appearance (Color)White
Infrared spectrumConforms
Melting point55°C to 64°C
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only