Carbon tetrabromide, 98%, Thermo Scientific Chemicals
Carbon tetrabromide, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Carbon tetrabromide, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
500 g
Packaging:
View all
Glass Bottle
Catalog number 325150050
Price (USD)/ Each
39.20
-
Add to cart
Quantity:
5 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)/ Each
39.20
Add to cart
Carbon tetrabromide, 98%, Thermo Scientific Chemicals
Catalog number325150050
Price (USD)/ Each
39.20
-
Add to cart
Chemical Identifiers
CAS558-13-4
IUPAC Nametetrabromomethane
Molecular FormulaCBr4
InChI KeyHJUGFYREWKUQJT-UHFFFAOYSA-N
SMILESBrC(Br)(Br)Br
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Water=<10 % (K.F.)
Appearance (Form)Crystalline solid
StabilizerMay contain water as stabilizer
GC>=97.5 % (On organic basis)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent
RUO – Research Use Only