Methanol-d4, for NMR, contains 0.03 v/v% TMS, 99.8% atom% D, AcroSeal™, Thermo Scientific Chemicals
Methanol-d4, for NMR, contains 0.03 v/v% TMS, 99.8% atom% D, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methanol-d4, for NMR, contains 0.03 v/v% TMS, 99.8% atom% D, AcroSeal™, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 325351000
Price (USD)/ Each
715.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
842.00 
Save 126.35 (15%)
-
Add to cart
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (USD)/ Each
715.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
842.00 
Save 126.35 (15%)
Add to cart
Methanol-d4, for NMR, contains 0.03 v/v% TMS, 99.8% atom% D, AcroSeal™, Thermo Scientific Chemicals
Catalog number325351000
Price (USD)/ Each
715.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
842.00 
Save 126.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Name(²H₃)methan(²H)ol
Molecular FormulaCH4O
InChI KeyOKKJLVBELUTLKV-MZCSYVLQSA-N
SMILES[2H]OC([2H])([2H])[2H]
Molecular Weight (g/mol)36.07
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
D-Enrichment>=99.75 %
TMSContains 0.03 v/v% TMS
Appearance (Form)Liquid
Water=<0.05 % (Coulometric)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For NMR
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only