Methyl sulfoxide-d6, for NMR, packaged in 0.50 ml ampoules, 99.9 atom % D, Thermo Scientific Chemicals
Methyl sulfoxide-d6, for NMR, packaged in 0.50 ml ampoules, 99.9 atom % D, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methyl sulfoxide-d6, for NMR, packaged in 0.50 ml ampoules, 99.9 atom % D, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
5 mL
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 325370050
Price (USD)/ Each
-
Quantity:
5 mL
Request bulk or custom format
Chemical Identifiers
IUPAC Name(²H₃)methanesulfinyl(²H₃)methane
Molecular FormulaC2H6OS
InChI KeyIAZDPXIOMUYVGZ-WFGJKAKNSA-N
SMILES[2H]C([2H])([2H])S(=O)C([2H])([2H])[2H]
Molecular Weight (g/mol)84.17
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Appearance (Form)Liquid
Water=<0.03 % (Coulometric)
D-Enrichment>=99.85 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For NMR
RUO – Research Use Only