Urethane, 97%
Urethane, 97%
Urethane, 97%
Thermo Scientific Chemicals

Urethane, 97%

CAS: 51-79-6 | C3H7NO2 | 89.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1 kg
Packaging:
Plastic bottle
Catalog number 325542500
Price (USD)
105.00
Each
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
105.00
Each
Add to cart
Chemical Identifiers
CAS51-79-6
IUPAC Nameethyl carbamate
Molecular FormulaC3H7NO2
InChI KeyJOYRKODLDBILNP-UHFFFAOYSA-N
SMILESCCOC(N)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals and/or chunks
Melting point46°C to 51°C
Infrared spectrumConforms
Appearance (Color)White to off-white
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only

Share catalog number, name or link