1,3,6,8-Pyrenetetrasulfonic acid tetrasodium salt, 85%, Thermo Scientific Chemicals
1,3,6,8-Pyrenetetrasulfonic acid tetrasodium salt, 85%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,3,6,8-Pyrenetetrasulfonic acid tetrasodium salt, 85%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 325560010
Price (USD)/ Each
91.70
-
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
91.70
Add to cart
1,3,6,8-Pyrenetetrasulfonic acid tetrasodium salt, 85%, Thermo Scientific Chemicals
Catalog number325560010
Price (USD)/ Each
91.70
-
Add to cart
Chemical Identifiers
CAS59572-10-0
IUPAC Nametetrasodium pyrene-1,3,6,8-tetrasulfonate
Molecular FormulaC16H6Na4O12S4
InChI KeyUZBIRLJMURQVMX-UHFFFAOYSA-J
SMILES[Na+].[Na+].[Na+].[Na+].[O-]S(=O)(=O)C1=CC(=C2C=CC3=C(C=C(C4=CC=C1C2=C34)S([O-])(=O)=O)S([O-])(=O)=O)S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow to green
Infrared spectrumConforms
Appearance (Form)Crystalline powder
UV spectrophotometry>=94 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only