Diethyl 1,3-acetonedicarboxylate, 95%, Thermo Scientific Chemicals
Diethyl 1,3-acetonedicarboxylate, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Diethyl 1,3-acetonedicarboxylate, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
250 mL
Packaging:
Glass bottle
Catalog number 325630500
Price (USD)/ Each
-
Quantity:
50 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS105-50-0
IUPAC Name1,5-diethyl 3-oxopentanedioate
Molecular FormulaC9H14O5
InChI KeyZSANYRMTSBBUCA-UHFFFAOYSA-N
SMILESCCOC(=O)CC(=O)CC(=O)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Appearance (Color)Clear colorless to light yellow
GC>=94 %
Infrared spectrumConforms
Refractive index1.4380 to 1.4420 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only