Thermo Scientific Chemicals

Acetonitrile, HPLC Gradient Grade, Thermo Scientific Chemicals

Catalog number: 325730010
1 L, Glass Bottle, Each
Thermo Scientific Chemicals

Acetonitrile, HPLC Gradient Grade, Thermo Scientific Chemicals

Catalog number: 325730010
1 L, Glass Bottle, Each
Quantity
Packaging
Catalog number: 325730010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
75-05-8
IUPAC Name
acetonitrile
Molecular Formula
C2H3N
InChI Key
WEVYAHXRMPXWCK-UHFFFAOYSA-N
SMILES
CC#N
LC Suitab. Gradient elution
passes test
Gradient analysis
:
UV
at 190 nm A: =<1.00
UV
at 220 nm A: =<0.01
UV
at 230 nm A: =<0.005

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text