Methyle4-oxo-3-piperidinecarboxylate hydrochloride, 97%, Thermo Scientific Chemicals
Methyle4-oxo-3-piperidinecarboxylate hydrochloride, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methyle4-oxo-3-piperidinecarboxylate hydrochloride, 97%, Thermo Scientific Chemicals

Have Questions?
Catalog number 325760010
Price (USD)/ Each
-
Request bulk or custom format
Chemical Identifiers
IUPAC Name(3R)-3-(methoxycarbonyl)-4-oxopiperidin-1-ium
Molecular FormulaC7H12NO3
InChI KeyXBNGBGYWMXBQRX-RXMQYKEDSA-O
SMILESCOC(=O)[C@@H]1C[NH2+]CCC1=O
Molecular Weight (g/mol)158.18
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to light pink
Appearance (Form)Powder
Infrared spectrumConforms
Titration Argentometric>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only