Thermo Scientific Chemicals

3-Bromopropionic acid, 97%, Thermo Scientific Chemicals

Catalog number: 325771000
100 g, Plastic bottle, Each
Thermo Scientific Chemicals

3-Bromopropionic acid, 97%, Thermo Scientific Chemicals

Catalog number: 325771000
100 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 325771000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
590-92-1
IUPAC Name
3-bromopropanoic acid
Molecular Formula
C3H5BrO2
InChI Key
DHXNZYCXMFBMHE-UHFFFAOYSA-N
SMILES
OC(=O)CCBr
Appearance (Color)
White to light yellow to orange
Appearance (Form)
Powder or crystals or flakes
Infrared spectrum
Conforms
Melting point
57°C to 63°C
HPLC
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text