Chemical Identifiers
CAS10378-47-9
IUPAC Nameλ⁴-cerium(4+) tetraamine dihydrate tetrahydrogen sulfate
Molecular FormulaCeH20N4O18S4
InChI KeyVCNAMBGKEDPVGQ-UHFFFAOYSA-J
SMILESN.N.N.N.O.O.[Ce+4].OS([O-])(=O)=O.OS([O-])(=O)=O.OS([O-])(=O)=O.OS([O-])(=O)=O
View more