Tetrahydropyran, 99%, Thermo Scientific Chemicals
Tetrahydropyran, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tetrahydropyran, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
Catalog number 325881000
Price (USD)/ Each
60.65
Online exclusive
67.20 
Save 6.55 (10%)
-
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
60.65
Online exclusive
67.20 
Save 6.55 (10%)
Add to cart
Tetrahydropyran, 99%, Thermo Scientific Chemicals
Catalog number325881000
Price (USD)/ Each
60.65
Online exclusive
67.20 
Save 6.55 (10%)
-
Add to cart
Chemical Identifiers
CAS142-68-7
IUPAC Nameoxane
Molecular FormulaC5H10O
InChI KeyDHXVGJBLRPWPCS-UHFFFAOYSA-N
SMILESC1CCOCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=98.5 %
Refractive index1.4190 to 1.4220 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only