3,4-Diaminothiophene dihydrochloride, 96%, Thermo Scientific Chemicals
3,4-Diaminothiophene dihydrochloride, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3,4-Diaminothiophene dihydrochloride, 96%, Thermo Scientific Chemicals

Have Questions?
Quantity:
1 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 325890010
Price (USD)/ Each
62.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
74.00 
Save 11.35 (15%)
-
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
62.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
74.00 
Save 11.35 (15%)
Add to cart
3,4-Diaminothiophene dihydrochloride, 96%, Thermo Scientific Chemicals
Catalog number325890010
Price (USD)/ Each
62.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
74.00 
Save 11.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Namethiophene-3,4-diamine dihydrochloride
Molecular FormulaC4H8Cl2N2S
InChI KeyRAMOMCXNLLLICQ-UHFFFAOYSA-N
SMILESCl.Cl.NC1=CSC=C1N
Molecular Weight (g/mol)187.08
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Grey to brown
Appearance (Form)Crystalline powder
Infrared spectrumConforms
HPLC>=95 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only