Thermo Scientific Chemicals

Hydroxylamine-O-sulfonic acid, 97%, Thermo Scientific Chemicals

Catalog number: 325940250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Hydroxylamine-O-sulfonic acid, 97%, Thermo Scientific Chemicals

Catalog number: 325940250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 325940250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2950-43-8
IUPAC Name
(aminooxy)sulfonic acid
Molecular Formula
H3NO4S
InChI Key
DQPBABKTKYNPMH-UHFFFAOYSA-N
SMILES
NOS(O)(=O)=O
Infrared spectrum
Conforms
Titration with NaOH
>=96.0 %
Appearance (Color)
White to off-white
Appearance (Form)
Crystalline powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text