Hydroxylamine-O-sulfonic acid, 97%, Thermo Scientific Chemicals
Hydroxylamine-O-sulfonic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Hydroxylamine-O-sulfonic acid, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
1 kg
Packaging:
Glass bottle
Catalog number 325941000
Price (USD)/ Each
232.65
Online exclusive
274.00 
Save 41.35 (15%)
-
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
232.65
Online exclusive
274.00 
Save 41.35 (15%)
Add to cart
Hydroxylamine-O-sulfonic acid, 97%, Thermo Scientific Chemicals
Catalog number325941000
Price (USD)/ Each
232.65
Online exclusive
274.00 
Save 41.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Name(aminooxy)sulfonic acid
Molecular FormulaH3NO4S
InChI KeyDQPBABKTKYNPMH-UHFFFAOYSA-N
SMILESNOS(O)(=O)=O
Molecular Weight (g/mol)113.09
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Titration with NaOH>=96.0 %
Appearance (Color)White to off-white
Appearance (Form)Crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only