Thermo Scientific Chemicals

Ethyl 2-methylacetoacetate, 95%, Thermo Scientific Chemicals

Catalog number: 326111000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Ethyl 2-methylacetoacetate, 95%, Thermo Scientific Chemicals

Catalog number: 326111000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 326111000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
609-14-3
IUPAC Name
ethyl 2-methyl-3-oxobutanoate
Molecular Formula
C7H12O3
InChI Key
FNENWZWNOPCZGK-UHFFFAOYNA-N
SMILES
CCOC(=O)C(C)C(C)=O
Water
=<1 % (K.F.)
Refractive index
1.4170 to 1.4230 (20°C, 589 nm)
Appearance (Form)
Liquid
Appearance (Color)
Clear colorless to yellow
GC
>=94 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text