Thermo Scientific Chemicals

3,4-Dichlorobenzyl bromide, 97%, Thermo Scientific Chemicals

Catalog number: 326161000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

3,4-Dichlorobenzyl bromide, 97%, Thermo Scientific Chemicals

Catalog number: 326161000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 326161000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
18880-04-1
IUPAC Name
4-(bromomethyl)-1,2-dichlorobenzene
Molecular Formula
C7H5BrCl2
InChI Key
XLWSBDFQAJXCQX-UHFFFAOYSA-N
SMILES
ClC1=C(Cl)C=C(CBr)C=C1
Infrared spectrum
Conforms
Appearance (Color)
Clear colorless to brown
Appearance (Form)
Liquid
Refractive index
1.6050 to 1.6080 (20°C, 589 nm)
GC
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text