Thermo Scientific Chemicals

Methyl 4-amino-3-chlorobenzoate, 97%, Thermo Scientific Chemicals

Catalog number: 326390050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

Methyl 4-amino-3-chlorobenzoate, 97%, Thermo Scientific Chemicals

Catalog number: 326390050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 326390050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
84228-44-4
IUPAC Name
methyl 4-amino-3-chlorobenzoate
Molecular Formula
C8H8ClNO2
InChI Key
WKVJBYKFGNVWLM-UHFFFAOYSA-N
SMILES
COC(=O)C1=CC=C(N)C(Cl)=C1
Infrared spectrum
Conforms
Appearance (Color)
Beige to light brown
HPLC
>=96.0 %
Appearance (Form)
Crystalline powder or crystals
Melting point
106°C to 110°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text