Dichloromethane, for residue and pestic. anal., stab. with amylene, Thermo Scientific Chemicals
Dichloromethane, for residue and pestic. anal., stab. with amylene, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Dichloromethane, for residue and pestic. anal., stab. with amylene, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
4 L
1 L
2.5 L
Catalog number 326600040
Price (USD)/ Each
210.65
Online exclusive
234.00 
Save 23.35 (10%)
-
Add to cart
Quantity:
4 L
Request bulk or custom format
Price (USD)/ Each
210.65
Online exclusive
234.00 
Save 23.35 (10%)
Add to cart
Dichloromethane, for residue and pestic. anal., stab. with amylene, Thermo Scientific Chemicals
Catalog number326600040
Price (USD)/ Each
210.65
Online exclusive
234.00 
Save 23.35 (10%)
-
Add to cart
Chemical Identifiers
CAS75-09-2
IUPAC Namedichloromethane
Molecular FormulaCH2Cl2
InChI KeyYMWUJEATGCHHMB-UHFFFAOYSA-N
SMILESClCCl
View more
SpecificationsSpecification SheetSpecification Sheet
Water=<0.02 % (K.F.)
GC(ECD) Passes test
StabilizerApprox. 50 ppm amylene
Appearance (Color)Clear colorless
Residue after evaporation=<2 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For residue analysis
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only