Thermo Scientific Chemicals

Ethyl acetate, for residue analysis, suitable for residue and pesticides analysis, Thermo Scientific Chemicals

Catalog number: 326610025
2.5 L, Glass Bottle, Each
Thermo Scientific Chemicals

Ethyl acetate, for residue analysis, suitable for residue and pesticides analysis, Thermo Scientific Chemicals

Catalog number: 326610025
2.5 L, Glass Bottle, Each
Quantity
Packaging
Catalog number: 326610025
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
141-78-6
IUPAC Name
ethyl acetate
Molecular Formula
C4H8O2
InChI Key
XEKOWRVHYACXOJ-UHFFFAOYSA-N
SMILES
CCOC(C)=O
Appearance (Form)
Liquid
Residue after evaporation
=<2 ppm
Appearance (Color)
Clear colorless
Water
=<0.05 % (Coulometric)
GC
(ECD) Passes test

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For residue analysis
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text