Water, for residue analysis, ECD tested for pesticide analysis, Thermo Scientific Chemicals
Water, for residue analysis, ECD tested for pesticide analysis, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Water, for residue analysis, ECD tested for pesticide analysis, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
Catalog number 326650010
Price (USD)/ Each
54.50
-
Add to cart
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)/ Each
54.50
Add to cart
Water, for residue analysis, ECD tested for pesticide analysis, Thermo Scientific Chemicals
Catalog number326650010
Price (USD)/ Each
54.50
-
Add to cart
Chemical Identifiers
CAS7732-18-5
IUPAC Namewater
Molecular FormulaH2O
InChI KeyXLYOFNOQVPJJNP-UHFFFAOYSA-N
SMILESO
View more
SpecificationsSpecification SheetSpecification Sheet
Acidity (CH3COOH)=<0.002 %
Residue after evaporation=<0.0002 %
Color scale=<10 APHA
Additional inforesidue (at bottling) : < 0.0005%
GC/ECD (as lindane)=<5 ng/l
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only