Thermo Scientific Chemicals

n-Heptane, 99+%, for residue analysis, ECD tested for pesticide analysis, Thermo Scientific Chemicals

Catalog number: 326690025
2.5 L, Each
Thermo Scientific Chemicals

n-Heptane, 99+%, for residue analysis, ECD tested for pesticide analysis, Thermo Scientific Chemicals

Catalog number: 326690025
2.5 L, Each
Quantity
Catalog number: 326690025
Price (USD)
Have Questions?

Chemical Identifiers

CAS
142-82-5
IUPAC Name
heptane
Molecular Formula
C7H16
InChI Key
IMNFDUFMRHMDMM-UHFFFAOYSA-N
SMILES
CCCCCCC
Appearance (Form)
Clear liquid
Color scale
=<10 APHA
GC
>=99.0 %
Acidity (CH3COOH)
=<0.002 %
Water
=<0.01 % (Coulometric)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For residue analysis
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text