Search Thermo Fisher Scientific
Danger
H319 - Causes serious eye irritation
H336 - May cause drowsiness or dizziness
H225 - Highly flammable liquid and vapor
EUH066 - Repeated exposure may cause skin dryness or cracking
P210 - Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking
P280 - Wear eye protection/ face protection
P303 + P361 + P353 - IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water or shower
P304 + P340 - IF INHALED: Remove person to fresh air and keep comfortable for breathing
P312 - Call a POISON CENTER or doctor if you feel unwell
P337 + P313 - If eye irritation persists: Get medical advice/attention