Dichloromethane, 99.9%, for residue analysis, for anal.of polyarom.hydrocarb., stab.with amylene, Thermo Scientific Chemicals
Dichloromethane, 99.9%, for residue analysis, for anal.of polyarom.hydrocarb., stab.with amylene, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Dichloromethane, 99.9%, for residue analysis, for anal.of polyarom.hydrocarb., stab.with amylene, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 L
1 L
2.5 L
Packaging:
View all
Metal Drum
Catalog number 326760100
Price (USD)/ Each
187.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
221.00 
Save 33.35 (15%)
-
Add to cart
Quantity:
10 L
Packaging:
Metal Drum
Request bulk or custom format
Price (USD)/ Each
187.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
221.00 
Save 33.35 (15%)
Add to cart
Dichloromethane, 99.9%, for residue analysis, for anal.of polyarom.hydrocarb., stab.with amylene, Thermo Scientific Chemicals
Catalog number326760100
Price (USD)/ Each
187.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
221.00 
Save 33.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Namedichloromethane
Molecular FormulaCH2Cl2
InChI KeyYMWUJEATGCHHMB-UHFFFAOYSA-N
SMILESClCCl
Molecular Weight (g/mol)84.93
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Water=<0.02 % (Coulometric)
UVat 240 nm A: =<0.108
Color scale=<5 APHA
UVat 235 nm A: =<0.5
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For residue analysis
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only