n-Heptane, 99+%, residue analysis, for trace analysis of polyaromatic hydrocarb, Thermo Scientific Chemicals
n-Heptane, 99+%, residue analysis, for trace analysis of polyaromatic hydrocarb, Thermo Scientific Chemicals
Thermo Scientific Chemicals

n-Heptane, 99+%, residue analysis, for trace analysis of polyaromatic hydrocarb, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 326770025
Price (USD)/ Each
-
Quantity:
2.5 L
Request bulk or custom format
Chemical Identifiers
IUPAC Nameheptane
Molecular FormulaC7H16
InChI KeyIMNFDUFMRHMDMM-UHFFFAOYSA-N
SMILESCCCCCCC
Molecular Weight (g/mol)100.21
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<10 APHA
Water=<0.001 % (Coulometric)
Residue after evaporation=<0.0003 %
GC>=99 %
Acidity=<0.002 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For residue analysis
RUO – Research Use Only