n-Hexane, 95%, for residue analysis, for trace analysis of polyaromatic hydrocarbons, Thermo Scientific Chemicals
n-Hexane, 95%, for residue analysis, for trace analysis of polyaromatic hydrocarbons, Thermo Scientific Chemicals
Thermo Scientific Chemicals

n-Hexane, 95%, for residue analysis, for trace analysis of polyaromatic hydrocarbons, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Packaging:
Glass Bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 326780025
Price (USD)/ Each
139.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
164.00 
Save 24.35 (15%)
-
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)/ Each
139.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
164.00 
Save 24.35 (15%)
Add to cart
n-Hexane, 95%, for residue analysis, for trace analysis of polyaromatic hydrocarbons, Thermo Scientific Chemicals
Catalog number326780025
Price (USD)/ Each
139.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
164.00 
Save 24.35 (15%)
-
Add to cart
Chemical Identifiers
CAS110-54-3
IUPAC Namehexane
Molecular FormulaC6H14
InChI KeyVLKZOEOYAKHREP-UHFFFAOYSA-N
SMILESCCCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<5 APHA
UVat 220 nm A: =<0.097
PAH (Poly Arom. Hydrocarbons)=<0.04 ppm
Appearance (Form)Clear liquid
GC>=95 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For residue analysis
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only