Acetonitrile, 99.9+%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
Acetonitrile, 99.9+%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Acetonitrile, 99.9+%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals

Acetonitrile, >-99.9%, C2H3N, CAS Number-75-05-8, methylkyanid, methane, cyano, methyl cyanide, mecn, methanecarbonitrile, ethanenitrile, ethyl nitrile, cyanure de methyl, cyanomethane, acetonitrile, 100mL, 0.002 % max., Vapor Pressure: 97mbar at 20 deg.C, 99.9+%, 02, 183, 81.0 deg.C to 82.0 deg.C
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
100 mL
1 L
Catalog number 326810025
Price (USD)/ Each
239.65
Online exclusive
282.00 
Save 42.35 (15%)
-
Add to cart
Quantity:
2.5 L
Request bulk or custom format
Price (USD)/ Each
239.65
Online exclusive
282.00 
Save 42.35 (15%)
Add to cart
Acetonitrile, 99.9+%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
Catalog number326810025
Price (USD)/ Each
239.65
Online exclusive
282.00 
Save 42.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Nameacetonitrile
Molecular FormulaC2H3N
InChI KeyWEVYAHXRMPXWCK-UHFFFAOYSA-N
SMILESCC#N
Molecular Weight (g/mol)41.05
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Residue after evaporation=<0.0005 %
Infrared spectrumConforms
Water=<0.001 % (Coulometric)
Acetamide=<10 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only