Thermo Scientific Chemicals

Cyclohexane, 99.5%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 326831000
100 mL, AcroSeal™ Glass Bottle, Each
Thermo Scientific Chemicals

Cyclohexane, 99.5%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 326831000
100 mL, AcroSeal™ Glass Bottle, Each
Quantity
Packaging
Catalog number: 326831000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
110-82-7
IUPAC Name
cyclohexane
Molecular Formula
C6H12
InChI Key
XDTMQSROBMDMFD-UHFFFAOYSA-N
SMILES
C1CCCCC1
Appearance (Color)
Clear colorless
Appearance (Form)
Liquid
Infrared spectrum
Conforms
GC
>=99.4 %
Water
=<0.005 % (Coulometric)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text