Thermo Scientific Chemicals

Dichloromethane, 99.9%, Extra Dry, stabilized, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 326850025
2.5 L, Each
Thermo Scientific Chemicals

Dichloromethane, 99.9%, Extra Dry, stabilized, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 326850025
2.5 L, Each
Quantity
Catalog number: 326850025
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
75-09-2
IUPAC Name
dichloromethane
Molecular Formula
CH2Cl2
InChI Key
YMWUJEATGCHHMB-UHFFFAOYSA-N
SMILES
ClCCl
Appearance (Color)
Clear colorless
Appearance (Form)
Liquid
Infrared spectrum
Conforms
GC
>=99.85 %
Water
=<0.003 % (Coulometric)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text