n-Hexane, 96+%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
n-Hexane, 96+%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals

n-Hexane, 96+%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 L
2.5 L
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 326921000
Price (USD)/ Each
60.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
71.00 
Save 10.35 (15%)
-
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (USD)/ Each
60.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
71.00 
Save 10.35 (15%)
Add to cart
n-Hexane, 96+%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
Catalog number326921000
Price (USD)/ Each
60.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
71.00 
Save 10.35 (15%)
-
Add to cart
Chemical Identifiers
CAS110-54-3
IUPAC Namehexane
Molecular FormulaC6H14
InChI KeyVLKZOEOYAKHREP-UHFFFAOYSA-N
SMILESCCCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Appearance (Form)Liquid
Filt.over 0.2 micron PFTEFiltered
GC>=96.0 %
Water=<0.002 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only