1-Methyl-2-pyrrolidinone, 99.5%, Extra Dry, AcroSeal™
1-Methyl-2-pyrrolidinone, 99.5%, Extra Dry, AcroSeal™
1-Methyl-2-pyrrolidinone, 99.5%, Extra Dry, AcroSeal™
1-Methyl-2-pyrrolidinone, 99.5%, Extra Dry, AcroSeal™
1-Methyl-2-pyrrolidinone, 99.5%, Extra Dry, AcroSeal™
Thermo Scientific Chemicals

1-Methyl-2-pyrrolidinone, 99.5%, Extra Dry, AcroSeal™

CAS: 872-50-4 | C5H9NO | 99.133 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
100 mL
1 L
Packaging:
View all
Glass Bottle
Catalog number 326930025
Price (USD)
294.65
Special offer
Online exclusive
Ends: 26-Sep-2025
346.00
Save 51.35 (15%)
Each
In stock
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)
294.65
Special offer
Online exclusive
Ends: 26-Sep-2025
346.00
Save 51.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS872-50-4
IUPAC Name1-methylpyrrolidin-2-one
Molecular FormulaC5H9NO
InChI KeySECXISVLQFMRJM-UHFFFAOYSA-N
SMILESCN1CCCC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Color scale=<20 APHA
GC>=99.4 %
Water=<0.005 %
Infrared spectrumConforms

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection