Methanol, 99.9%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
Methanol, 99.9%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methanol, 99.9%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals

Methanol, 99.9%, CH4O, CAS Number-67-56-1, methylol, methyl alcohol, colonial spirit, carbinol, wood alcohol, wood naphtha, pyroxylic spirit, methyl hydroxide, wood spirit, columbian spirit, 1L, 0.002% max. (as HCOOH), 0.0005 meq/g max., 64.7 deg.C, CHEBI:17790, Colorless, 0.7910g/mL, 200-659-6
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
100 mL
1 L
Packaging:
View all
Glass Bottle
Catalog number 326950025
Price (USD)/ Each
108.65
Online exclusive
121.00 
Save 12.35 (10%)
-
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)/ Each
108.65
Online exclusive
121.00 
Save 12.35 (10%)
Add to cart
Methanol, 99.9%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
Catalog number326950025
Price (USD)/ Each
108.65
Online exclusive
121.00 
Save 12.35 (10%)
-
Add to cart
Chemical Identifiers
CAS67-56-1
IUPAC Namemethanol
Molecular FormulaCH4O
InChI KeyOKKJLVBELUTLKV-UHFFFAOYSA-N
SMILESCO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=99.85 %
Water=<0.005 % (Coulometric)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only