Isopropanol, 99.8%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
Isopropanol, 99.8%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Isopropanol, 99.8%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
100 mL
2.5 L
Catalog number 326960010
Price (USD)/ Each
71.65
Online exclusive
79.80 
Save 8.15 (10%)
-
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (USD)/ Each
71.65
Online exclusive
79.80 
Save 8.15 (10%)
Add to cart
Isopropanol, 99.8%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
Catalog number326960010
Price (USD)/ Each
71.65
Online exclusive
79.80 
Save 8.15 (10%)
-
Add to cart
Chemical Identifiers
CAS67-63-0
IUPAC Namepropan-2-ol
Molecular FormulaC3H8O
InChI KeyKFZMGEQAYNKOFK-UHFFFAOYSA-N
SMILESCC(C)O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)Clear colorless
Appearance (Form)Liquid
GC>=99.75 %
Water=<0.005 % (Coulometric)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only