Thermo Scientific Chemicals

Tetrahydrofuran, 99.85%, Extra Dry, stabilized, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 326970010
1 L, Glass Bottle, Each
Thermo Scientific Chemicals

Tetrahydrofuran, 99.85%, Extra Dry, stabilized, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 326970010
1 L, Glass Bottle, Each
Quantity
Packaging
Catalog number: 326970010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
109-99-9
IUPAC Name
oxolane
Molecular Formula
C4H8O
InChI Key
WYURNTSHIVDZCO-UHFFFAOYSA-N
SMILES
C1CCOC1
Appearance (Color)
Clear colorless
Appearance (Form)
Liquid
GC
>=99.80 %
Water
=<0.005 % (Coulometric)
Acidity (CH3COOH)
=<0.0005 meq/g

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text