Tetrahydrofuran, 99.85%, Extra Dry, stabilized, AcroSeal™, Thermo Scientific Chemicals
Tetrahydrofuran, 99.85%, Extra Dry, stabilized, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tetrahydrofuran, 99.85%, Extra Dry, stabilized, AcroSeal™, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 L
2.5 L
Packaging:
View all
AcroSeal™ Glass Bottle
Catalog number 326971000
Price (USD)/ Each
65.65
Online exclusive
72.60 
Save 6.95 (10%)
-
Add to cart
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (USD)/ Each
65.65
Online exclusive
72.60 
Save 6.95 (10%)
Add to cart
Tetrahydrofuran, 99.85%, Extra Dry, stabilized, AcroSeal™, Thermo Scientific Chemicals
Catalog number326971000
Price (USD)/ Each
65.65
Online exclusive
72.60 
Save 6.95 (10%)
-
Add to cart
Chemical Identifiers
IUPAC Nameoxolane
Molecular FormulaC4H8O
InChI KeyWYURNTSHIVDZCO-UHFFFAOYSA-N
SMILESC1CCOC1
Molecular Weight (g/mol)72.11
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Appearance (Form)Liquid
GC>=99.80 %
Water=<0.005 % (Coulometric)
Acidity (CH3COOH)=<0.0005 meq/g
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only