Search Thermo Fisher Scientific
General description
• Phenol/Tris is a solvent system consisting of phenol and Tris(Hydroxymethyl)aminomethane
• Prepared as ready-to-use, no titration necessary
Applications
• Commonly used in DNA isolation. Tris(Hydroxymethyl)aminomethane acts as a proton acceptor to raise pH