Boric acid, 99.5%, for molecular biology, DNAse, RNAse and Protease free, Thermo Scientific Chemicals
Boric acid, 99.5%, for molecular biology, DNAse, RNAse and Protease free, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Boric acid, 99.5%, for molecular biology, DNAse, RNAse and Protease free, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
Catalog number 327130010
Price (USD)/ Each
83.65
Online exclusive
92.40 
Save 8.75 (9%)
-
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (USD)/ Each
83.65
Online exclusive
92.40 
Save 8.75 (9%)
Add to cart
Boric acid, 99.5%, for molecular biology, DNAse, RNAse and Protease free, Thermo Scientific Chemicals
Catalog number327130010
Price (USD)/ Each
83.65
Online exclusive
92.40 
Save 8.75 (9%)
-
Add to cart
Chemical Identifiers
CAS10043-35-3
IUPAC Nameboric acid
Molecular FormulaBH3O3
InChI KeyKGBXLFKZBHKPEV-UHFFFAOYSA-N
SMILESOB(O)O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Powder
Dnase,Rnase,Protease activitynone detected
Chloride (Cl)=<10 ppm
Phosphate (PO4)=<10 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer, Lewis acid
RUO – Research Use Only