Thermo Scientific Chemicals

Citric acid, trisodium salt hydrate, 99.8%, for molecular biology, DNAse, RNAse and, Thermo Scientific Chemicals

Catalog number: 327160010
1 kg, Glass Bottle, Each
Thermo Scientific Chemicals

Citric acid, trisodium salt hydrate, 99.8%, for molecular biology, DNAse, RNAse and, Thermo Scientific Chemicals

Catalog number: 327160010
1 kg, Glass Bottle, Each
Quantity
Packaging
Catalog number: 327160010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
114456-61-0
IUPAC Name
trisodium 2-hydroxypropane-1,2,3-tricarboxylate
Molecular Formula
C6H5Na3O7
InChI Key
HRXKRNGNAMMEHJ-UHFFFAOYSA-K
SMILES
[Na+].[Na+].[Na+].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O
Infrared spectrum
Conforms
Appearance (Color)
White
Appearance (Form)
Crystalline powder or crystals
Assay
>=99.75 % (on dry substance)
Loss on drying
12 % to 13 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text