Thermo Scientific Chemicals

DL-1,4-Dithiothreitol, 99+%, for molecular biology, DNAse, RNAse and Protease free

Catalog number: 327190100
10 g, Plastic Bottle, Each
Thermo Scientific Chemicals

DL-1,4-Dithiothreitol, 99+%, for molecular biology, DNAse, RNAse and Protease free

Catalog number: 327190100
10 g, Plastic Bottle, Each
Quantity
Packaging
Catalog number: 327190100
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3483-12-3
IUPAC Name
(2R,3R)-1,4-disulfanylbutane-2,3-diol
Molecular Formula
C4H10O2S2
InChI Key
VHJLVAABSRFDPM-IMJSIDKUSA-N
SMILES
O[C@@H](CS)[C@@H](O)CS
UV
at 280 nm A: =<0.1
Appearance (Color)
White to off-white
Infrared spectrum
Conforms
Titration Iodometric
99.0 to 101.0 %
Heavy metals (as Pb)
=<5 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text