Thermo Scientific Chemicals

HEPES sodium salt, 99+%, for molecular biology, DNAse, RNAse and Protease free

Catalog number: 327265000
500 g, Plastic bottle, Each
Thermo Scientific Chemicals

HEPES sodium salt, 99+%, for molecular biology, DNAse, RNAse and Protease free

Catalog number: 327265000
500 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 327265000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
75277-39-3
IUPAC Name
sodium 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethane-1-sulfonate
Molecular Formula
C8H17N2NaO4S
InChI Key
RDZTWEVXRGYCFV-UHFFFAOYSA-M
SMILES
[Na+].OCCN1CCN(CCS([O-])(=O)=O)CC1
Appearance (Form)
Crystalline powder
UV
at 280 nm A: =<0.02
Appearance (Color)
White
Infrared spectrum
Conforms
Titration with HClO4
>=99.0 % (on dry substance)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text