Sodium chloride, 99.85%, for molecular biology, DNAse, RNAse and Protease free
Sodium chloride, 99.85%, for molecular biology, DNAse, RNAse and Protease free
Thermo Scientific Chemicals

Sodium chloride, 99.85%, for molecular biology, DNAse, RNAse and Protease free

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
2.5 kg
Catalog number 327300010
Price (USD)/ Each
88.65
Online exclusive
98.80 
Save 10.15 (10%)
-
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (USD)/ Each
88.65
Online exclusive
98.80 
Save 10.15 (10%)
Add to cart
Sodium chloride, 99.85%, for molecular biology, DNAse, RNAse and Protease free
Catalog number327300010
Price (USD)/ Each
88.65
Online exclusive
98.80 
Save 10.15 (10%)
-
Add to cart
Chemical Identifiers
CAS7647-14-5
IUPAC Namesodium chloride
Molecular FormulaClNa
InChI KeyFAPWRFPIFSIZLT-UHFFFAOYSA-M
SMILES[Na+].[Cl-]
View more
SpecificationsSpecification SheetSpecification Sheet
UVat 280 nm A: =<0.01
Sulfate (SO4)=<4 ppm
Iron (Fe)=<2 ppm
Calcium (Ca)=<50 ppm
Magnesium (Mg)=<50 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer
RUO – Research Use Only