Dodecyl sulfate sodium salt, 99+%, for molecular biology, DNAse, RNAse and Protease free
Dodecyl sulfate sodium salt, 99+%, for molecular biology, DNAse, RNAse and Protease free
Thermo Scientific Chemicals

Dodecyl sulfate sodium salt, 99+%, for molecular biology, DNAse, RNAse and Protease free

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
100 g
500 g
Catalog number 327310025
Price (USD)/ Each
-
Quantity:
2.5 kg
Request bulk or custom format
Chemical Identifiers
CAS151-21-3, 7757-82-6, 7647-14-5
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay>=99.0 %
Chloride (Cl)=<1000 ppm
UVat 260 nm A: =<0.05
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only