Thermo Scientific Chemicals

Tris(hydroxymethyl)aminomethane, 99.85%, for molecular biology, DNAse, RNAse and Pro

Catalog number: 327360050
5 kg, Plastic Drum, Each
Thermo Scientific Chemicals

Tris(hydroxymethyl)aminomethane, 99.85%, for molecular biology, DNAse, RNAse and Pro

Catalog number: 327360050
5 kg, Plastic Drum, Each
Quantity
Packaging
Catalog number: 327360050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
77-86-1
IUPAC Name
2-amino-2-(hydroxymethyl)propane-1,3-diol
Molecular Formula
C4H11NO3
InChI Key
LENZDBCJOHFCAS-UHFFFAOYSA-N
SMILES
NC(CO)(CO)CO
UV
at 260 nm A: =<0.03
Appearance (Form)
Crystalline powder or crystals
Water
=<1 %
Arsenic (As)
=<5 ppm
Iron (Fe)
=<1 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text