Sodium tert-pentoxide, 95%, pure, Thermo Scientific Chemicals
Sodium tert-pentoxide, 95%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sodium tert-pentoxide, 95%, pure, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
Catalog number 327551000
Price (USD)/ Each
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Namesodium 2,2-dimethylpropan-1-olate
Molecular FormulaC5H11NaO
InChI KeyYQMLGVNRTAQUFQ-UHFFFAOYSA-N
SMILES[Na+].CC(C)(C)C[O-]
Molecular Weight (g/mol)110.13
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Cream to tan
Titration with HClO4>=94 %
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only