Thermo Scientific Chemicals

Diisopropyl azodicarboxylate, 94%, Thermo Scientific Chemicals

Catalog number: 327561000
100 g, Each
Thermo Scientific Chemicals

Diisopropyl azodicarboxylate, 94%, Thermo Scientific Chemicals

Catalog number: 327561000
100 g, Each
Quantity
Catalog number: 327561000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2446-83-5
IUPAC Name
N-{[(propan-2-yloxy)carbonyl]imino}(propan-2-yloxy)formamide
Molecular Formula
C8H14N2O4
InChI Key
VVWRJUBEIPHGQF-UHFFFAOYSA-N
SMILES
CC(C)OC(=O)N=NC(=O)OC(C)C
Appearance (Color)
Clear to hazy yellow to orange to red
Infrared spectrum
Conforms
Refractive index
1.4180 to 1.4220 (20°C, 589 nm)
Appearance (Form)
Liquid
GC
>=92 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X Bond Formation (Halogen), Radical initiators
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text