ACES, 99+%, for biochemistry
ACES, 99+%, for biochemistry
Thermo Scientific Chemicals

ACES, 99+%, for biochemistry

ACES, >-99%, C4H10N2O4S, CAS Number-7365-82-4, N-(2-Acetamido)-2-aminoethanesulfonic acid, N-(Carbamoylmethyl)taurine, 1kg, 0.02 max. at 280nm, 0.04 max. at 260nm, (5% aq. soln.), (5% aq. soln.) at 0nm, 99+%, 100.0, 99.0, CHEBI:39062, 100ppm max., White, 230-908-4, 182.19, Biochemical
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
250 g
1 kg
Packaging:
Plastic bottle
Catalog number 327671000
Price (USD)/ Each
154.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
182.00 
Save 27.35 (15%)
-
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
154.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
182.00 
Save 27.35 (15%)
Add to cart
ACES, 99+%, for biochemistry
Catalog number327671000
Price (USD)/ Each
154.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
182.00 
Save 27.35 (15%)
-
Add to cart
Chemical Identifiers
CAS7365-82-4
IUPAC Name2-[(carbamoylmethyl)amino]ethane-1-sulfonic acid
Molecular FormulaC4H10N2O4S
InChI KeyDBXNUXBLKRLWFA-UHFFFAOYSA-N
SMILESNC(=O)CNCCS(O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Titration with NaOH>=99.0 %
pH3.6 to 4.4 (1 % at 25°C)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer for biochemistry
RUO – Research Use Only