Informational:Informational:This product ships via ground from US northeast.
Thermo Scientific Chemicals

2,2-Bis(hydroxymethyl)-2,2',2''-nitrilotriethanol, 99+%, for biochemistry

Catalog number: 327721000
100 g, Each
Thermo Scientific Chemicals

2,2-Bis(hydroxymethyl)-2,2',2''-nitrilotriethanol, 99+%, for biochemistry

Catalog number: 327721000
100 g, Each
Quantity
Catalog number: 327721000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6976-37-0
IUPAC Name
2-[bis(2-hydroxyethyl)amino]-2-(hydroxymethyl)propane-1,3-diol
Molecular Formula
C8H19NO5
InChI Key
OWMVSZAMULFTJU-UHFFFAOYSA-N
SMILES
OCCN(CCO)C(CO)(CO)CO
Appearance (Form)
Powder
Infrared spectrum
Conforms
Titration with HClO4
>=99.0 %
Water
=<1.0 % (K.F.)
Iron (Fe)
=<5 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text