Thermo Scientific Chemicals

CAPSO, 99%

Catalog number: 327741000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

CAPSO, 99%

Catalog number: 327741000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 327741000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
73463-39-5
IUPAC Name
(2S)-3-(cyclohexylazaniumyl)-2-hydroxypropane-1-sulfonate
Molecular Formula
C9H19NO4S
InChI Key
INEWUCPYEUEQTN-VIFPVBQESA-N
SMILES
O[C@@H](C[NH2+]C1CCCCC1)CS([O-])(=O)=O
Appearance (Color)
White
Appearance (Form)
Powder
Infrared spectrum
Conforms
Titration with NaOH
>=98.5 %
pH
5 to 6.5 (1 % in water at 25°C)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text